missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Arginase 2 Recombinant Rabbit Monoclonal Antibody (23GB655), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Arginase 2 |
|---|---|
| Clone | 23GB655 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089286
![]() |
Invitrogen™
MA554192 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089266
|
Invitrogen™
MA554193 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Arginase 2 Recombinant Monoclonal antibody specifically detects Arginase 2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Arginase 2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P78540 | |
| 384 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB655 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AII; AII type II arginase; ARG2; arginase 2; arginase II; arginase type II; arginase, type II; arginase-2, mitochondrial; AU022422; kidney arginase; kidney-type arginase; L-arginine amidinohydrolase; L-arginine ureahydrolase; nonhepatic arginase; non-hepatic arginase; type II arginase | |
| A synthetic peptide of human Arg2 (301-354AA). | |
| Primary | |
| -20°C | |
| ARG2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title