missing translation for 'onlineSavingsMsg'
Learn More

Arginase 1/ARG1/liver Arginase Chicken anti-Human, Mouse, Rat, Guinea Pig, Polyclonal, Novus Biologicals™

Product Code. 18346014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346014 100 μg 100µL
18322883 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18346014 Supplier Novus Biologicals Supplier No. NBP311945100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

Arginase 1/ARG1/liver Arginase Polyclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arginase 1/ARG1/liver Arginase
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:500
Formulation 100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol.
Gene Alias ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase
Host Species Chicken
Immunogen Full-length recombinant rat arginase I. (UniProt: P07824)
Purification Method IgY purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 383
Target Species Human, Mouse, Rat, Guinea Pig
Content And Storage Store at -20C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.