missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Chicken anti-Human, Mouse, Rat, Guinea Pig, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Arginase 1/ARG1/liver Arginase Polyclonal antibody specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse, Rat, Guinea Pig samples. It is validated for Western Blot, Immunohistochemistry
Specifications
Specifications
| Antigen | Arginase 1/ARG1/liver Arginase |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:500 |
| Formulation | 100 ul in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. |
| Gene Alias | ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver-type arginase, Type I arginase |
| Host Species | Chicken |
| Immunogen | Full-length recombinant rat arginase I. (UniProt: P07824) |
| Purification Method | IgY purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?