missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginase 1/ARG1/liver Arginase Antibody (ARG1/1125 + ARG1/1126), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £373.00
Specifications
| Antigen | Arginase 1/ARG1/liver Arginase |
|---|---|
| Clone | ARG1/1125 + ARG1/1126 |
| Concentration | 0.2mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Immunofluorescence 1 - 2 ug/ml |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18198778
|
Novus Biologicals
NBP2-44967-0.02MG |
0.02 mg |
£221.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18120408
|
Novus Biologicals
NBP2-44967-0.1MG |
0.1 mg |
£373.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Arginase 1/ARG1/liver Arginase | |
| 0.2mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase | |
| ARG1 | |
| IgG | |
| Protein A or G purified | |
| Recognizes a protein of 35-38kDa, which is identified as Arginase 1 (ARG1). Arginase is a manganese metallo-enzyme that catalyzes the hydrolysis of arginine to generate ornithine and urea. Arginase I and II are isoenzymes, which differ in subcellular localization, regulation, and possibly function. Arginase I is a cytosolic enzyme, which is expressed mainly in the liver as part of the urea cycle, whereas arginase II is a mitochondrial protein found in a variety of tissues. Antibody to ARG-1 labels hepatocytes in normal tissues and granulocytes in peripheral blood. ARG-1 is a sensitive and specific marker for identification of hepatocellular carcinoma. |
| ARG1/1125 + ARG1/1126 | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P05089 | |
| 383 | |
| Recombinant fragment (87 Amino acid residues around aa 1-150) of human ARG1 protein | |
| Primary | |
| Store at 4C. | |
| 36.5 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title