missing translation for 'onlineSavingsMsg'
Learn More

Arginase 1/ARG1/liver Arginase Antibody (034), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18380401 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380401 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380401 Supplier Novus Biologicals Supplier No. NBP289915AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Arginase 1/ARG1/liver Arginase Monoclonal specifically detects Arginase 1/ARG1/liver Arginase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arginase 1/ARG1/liver Arginase
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 34
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, Liver-type arginase, Type I arginase
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Arginase 1/ARG1/liver Arginase (P05089-1; Met 1-Lys 322).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Chromatin Research, Lipid and Metabolism, Myeloid derived Suppressor Cell
Primary or Secondary Primary
Gene ID (Entrez) 383
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.