missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Argatroban

Product Code. 18054132
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18054132 50 mg 50mg
18045932 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054132 Supplier R&D Systems™ Supplier No. 1637/50
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent thrombin inhibitor

Potent inhibitor of thrombin mediated fibrinogen cleavage (Ki = 19 nM). Competitive inhibitor of thrombin-induced platelet activation and clotting. Shown to exhibit antithrombotic activity in animal models.

This compound is a mixture of 2 diastereomers (approximate ratio 65/35).

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Argatroban
CAS 74863-84-6
Assay Percent Range >99%
Quantity 50 mg
Product Line Tocris
Molecular Formula C23H36N6O5S
Formula Weight 508.63
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.