missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARG1 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5D6D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66129-1-IG-150UL
This item is not returnable.
View return policy
Description
Arginase-1 (ARG1) catalyzes the hydrolysis of L-arginine into ornithine and urea, a critical step for the urea cycle. Defects in ARG1 are the cause of argininemia, an autosomal recessive disorder characterized by hyperammonemia. There are several isoforms of mammalian arginase. The type I isoform, is a cytosolic enzyme and expressed predomitly in the liver as a component of the urea cycle. Hepatocellular carcinoma usually shows higher protein expression of ARG1 than normal liver cells.Specifications
| ARG1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P05089, P07824, Q61176, Q95JC8 | |
| Arg1 | |
| ARG1 Fusion Protein Ag8810 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 5D6D12 | |
| Unconjugated | |
| ARG1 | |
| ARG1, Arginase 1, liver Arginase, Liver type arginase | |
| Mouse | |
| Protein G | |
| RUO | |
| 11846, 29221, 383, 397115 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction