Learn More
ARG1 Mouse anti-Human, Clone: ARG1/1126, Abnova™
Mouse monoclonal antibody raised against partial recombinant human ARG1.
Brand: Abnova MAB13326.100ug
Description
Arginase catalyzes the hydrolysis of arginine to ornithine and urea. At least two isoforms of mammalian arginase exist (types I and II) which differ in their tissue distribution, subcellular localization, immunologic crossreactivity and physiologic function. The type I isoform encoded by this gene, is a cytosolic enzyme and expressed predominantly in the liver as a component of the urea cycle. Inherited deficiency of this enzyme results in argininemia, an autosomal recessive disorder characterized by hyperammonemia. [provided by RefSeq]
Specifications
| ARG1 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| ARG1 | |
| Recombinant protein corresponding to 87 residues around amino acids 1-150 of human ARG1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| ARG1/1126 | |
| Mouse monoclonal antibody raised against partial recombinant human ARG1. | |
| In 10mM PBS. | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 383 | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG3 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.