Learn More
Eurofins DiscoverX Arg Human Protein, Active
Description
The mammalian Abelson (Abl) family of nonreceptor tyrosine kinases is comprised of c-Abl (ABL1) and Arg (Abl-related gene, ABL2), which are encoded by the Abl1 and Abl2 genes, respectively. Deregulation of the c-Abl and Arg tyrosine kinases can occur as a result of chromosomal translocation events that produce chimeric proteins associated with the development of human leukemias as well as gastric cancer invasion and progression.
GenBank NM_005158
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution. |
| Molecular Weight (g/mol) | 121.9 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Truncated |
| Purity or Quality Grade | ≥31% |
| Protein | Arg, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.