missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX Arg Human Protein, Active

Product Code. 17380284
Encompass_Preferred
Click to view available options
Quantity:
10 μg
250 μg
Unit Size:
10µg
250µg
This item is not returnable. View return policy

Product Code. 17380284

Brand: Eurofins DiscoverX 14521

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, human, N-terminal His6-tagged recombinant human Arg, residues 38-end, for use in Kinase Assays.

The mammalian Abelson (Abl) family of nonreceptor tyrosine kinases is comprised of c-Abl (ABL1) and Arg (Abl-related gene, ABL2), which are encoded by the Abl1 and Abl2 genes, respectively. Deregulation of the c-Abl and Arg tyrosine kinases can occur as a result of chromosomal translocation events that produce chimeric proteins associated with the development of human leukemias as well as gastric cancer invasion and progression.

GenBank NM_005158

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35. Frozen solution.
Molecular Weight (g/mol) 121.9 kDa
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥31%
Protein Arg, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.