missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Arg-Gly-Asp-Cys

Product Code. 16449478
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449478 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449478 Supplier MedChemExpress Supplier No. HYP0314
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Arg-Gly-Asp-Cys is the binding motif of fibronectin to cell adhesion molecules, and can inhibit platelet aggregation and fibrinogen binding.

Specifications

CAS 109292-46-8
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 449.48 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.