missing translation for 'onlineSavingsMsg'
Learn More

Arg-AMS, MedChemExpress

Product Code. 16483216
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483216 25 mg 25mg
16467437 5 mg 5mg
16424728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16483216 Supplier Medchem Express Supplier No. HY112862

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Arg-AMS is a potent nanomolar inhibitor of arginyl tRNA synthetase, which displays tightly bound inhibitory characteristics for the A-domains in non-ribosomal peptide synthetases (NRPS) enzymes.

Arg-AMS is a potent nanomolar inhibitor of arginyl tRNA synthetase, which displays tightly bound inhibitory characteristics for the A-domains in non-ribosomal peptide synthetases (NRPS) enzymes.

Specifications

Chemical Name or Material Arg-AMS
CAS 301351-95-1
Color White
Purity Grade Notes Research
Molecular Formula C16H26N10O7S
Quantity 25 mg
Solubility Information DMSO : ≥ 100 mg/mL (199.00 mM)
SMILES O[C@@H]([C@H]([C@H](N1C=NC2=C1N=CN=C2N)O3)O)[C@H]3COS(=O)(NC([C@@H](N)CCCNC(N)=N)=O)=O
Molecular Weight (g/mol) 502.51
Formula Weight 502.51
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.