missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARFRP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102967
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARFRP1.
The protein encoded by this gene is a membrane-associated GTP-ase which localizes to the plasma membrane and is related to the ADP-ribosylation factor (ARF) and ARF-like (ARL) proteins. This gene plays a role in membrane trafficking between the trans-Golgi network and endosomes. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ARFRP1 | |
| Polyclonal | |
| Unconjugated | |
| ARFRP1 | |
| 1500006I01Rik; ADP ribosylation factor related protein 1; ADP-ribosylation factor related protein 1; ADP-ribosylation factor-related protein 1; AI480700; ARF-related protein 1; Arfrp1; ARL18; ARP; Arp1; helicase-like protein NHL; SCG10 like-protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10139 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13795 | |
| ARFRP1 | |
| A synthesized peptide derived from human ARFRP1(Accession Q13795), corresponding to amino acid residues A133-M183. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction