missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFRP1 Antibody (SR1362), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22541-50ul
This item is not returnable.
View return policy
Description
ARFRP1 Monoclonal antibody specifically detects ARFRP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| ARFRP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1362 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| ADP-ribosylation factor related protein 1, ARF-related protein 1, ARL18, ARP1, ARPADP-ribosylation factor-related protein 1, helicase-like protein NHL, SCG10 like-protein | |
| A synthesized peptide derived from human ARFRP1 (Uniprot #: Q13795) | |
| 50 μg | |
| Signal Transduction | |
| 10139 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction