missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ARFIP2 |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16833244
|
Proteintech
14548-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16823244
|
Proteintech
14548-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Putative target protein of ADP-ribosylation factor. Involved in membrane ruffling.Specifications
| ARFIP2 | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| P53365, Q6AY65, Q8K221 | |
| 23647, 293344, 76932 | |
| ARFIP2 Fusion Protein Ag6076 | |
| Primary | |
| -20°C | |
| ARFIP2 |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Arfaptin 2, ARFIP2, Partner of RAC1, POR1, Protein POR1 | |
| Arfip2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title