missing translation for 'onlineSavingsMsg'
Learn More

ARFIP2 Antibody (2B5), Novus Biologicals™

Product Code. 18400130 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400130 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400130 Supplier Novus Biologicals Supplier No. H00023647M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARFIP2 Monoclonal antibody specifically detects ARFIP2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARFIP2
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 2B5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH00392
Gene Alias ADP-ribosylation factor interacting protein 2, ADP-ribosylation factor-interacting protein 2, arfaptin 2, arfaptin-2, Partner of RAC1, POR1partner of RAC1 (arfaptin 2), Protein POR1
Host Species Mouse
Immunogen ARFIP2 (AAH00392, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MTDGILGKAATMEIPIHGNGEARQLPEDDGLEQDLQQVMVSGPNLNETSIVSGGYGGSGDGLIPTGSGRHPSHSTTPSGPGDEVARGIAGEKFDIVKKWGINTYKCTKQL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23647
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.