missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARFGEF2 Recombinant Rabbit Monoclonal Antibody (24GB6840)

Rabbit Recombinant Monoclonal Antibody
£126.00 - £434.00
Specifications
| Antigen | ARFGEF2 |
|---|---|
| Clone | 24GB6840 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275363
![]() |
Invitrogen™
MA556969 |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274875
|
Invitrogen™
MA556970 |
20 μL |
£126.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARFGEF2 Recombinant Monoclonal Antibody for Western Blot
ADP-ribosylation factors (ARFs) play an important role in intracellular vesicular trafficking. The protein encoded by this gene is involved in the activation of ARFs by accelerating replacement of bound GDP with GTP and is involved in Golgi transport. It contains a Sec7 domain, which may be responsible for its guanine-nucleotide exchange activity and also brefeldin A inhibition. [provided by RefSeq, Jul 2008].Specifications
| ARFGEF2 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| ARFGEF2 | |
| ADP ribosylation factor guanine nucleotide exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2; ADP-ribosylation factor guanine nucleotide-exchange factor 2 (brefeldin A-inhibited); AI463430; Arfgef2; ARFGEP2; AW493672; Big2; Brefeldin A-inhibited GEP 2; Brefeldin A-inhibited guanine nucleotide-exchange factor 2; brefeldin A-inhibited guanine nucleotide-exchange protein 2; dJ1164I10.1; E230011G24Rik; PVNH2 | |
| ARFGEF2 | |
| Primary | |
| Affinity chromatography |
| 24GB6840 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2A5R2, Q7TSU1, Q9Y6D5 | |
| 10564, 296380, 99371 | |
| A synthesized peptide derived from human ARFGEF2 (aa 1501-1514). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title