missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFGEF1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-55257UV
This item is not returnable.
View return policy
Description
ARFGEF1 Polyclonal antibody specifically detects ARFGEF1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ARFGEF1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldinA-inhibited), ARFGEP1P200, BIG1p200, Brefeldin A-inhibited GEP 1, brefeldin A-inhibited guanine nucleotide-exchange protein 1, D730028O18Rik, DKFZP434L057, p200 ARF guanine nucleotide exchange factor, p200 ARF-GEP1 | |
| The immunogen recognized by this antibody maps to a region between residue 1800 and the C-terminus (residue 1849) of human brefeldin A-inhibited guanine nucleotide-exchange protein 1 using the numbering given in entry NP_006412.2 (GeneID 10565). | |
| 0.1 mL | |
| Signal Transduction | |
| 10565 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction