missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFGEF1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92320-0.02ml
This item is not returnable.
View return policy
Description
ARFGEF1 Polyclonal antibody specifically detects ARFGEF1 in Human samples. It is validated for Western Blot
Specifications
| ARFGEF1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldinA-inhibited), ARFGEP1P200, BIG1p200, Brefeldin A-inhibited GEP 1, brefeldin A-inhibited guanine nucleotide-exchange protein 1, D730028O18Rik, DKFZP434L057, p200 ARF guanine nucleotide exchange factor, p200 ARF-GEP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-420 of human ARFGEF1 (NP_006412.2). MEKERHRQHHHLLQSPVSHHEPESPQLRYLPPQTVDHISQEHEGDLDLHTNDVDKSLQDDTEPENGSDISSAENEQTEADQATAAETLSKNEVLYDGENHDCEEKPQDIVQNIVEEMVNIVVGDMGEGTTINASADGNIGTIEDGSDSENIQANGIPGTPISVAYTPSLPDDRLSVSSNDTQESGNSSGPSPGAKFSHILQ | |
| 0.02 mL | |
| Signal Transduction | |
| 10565 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction