missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFGAP3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15442
This item is not returnable.
View return policy
Description
ARFGAP3 Polyclonal specifically detects ARFGAP3 in Human samples. It is validated for Western Blot.
Specifications
| ARFGAP3 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase activating protein 3, ADP-ribosylation factor GTPase-activating protein 3, ARF GAP 3, ARFGAP1, FLJ45618 | |
| Rabbit | |
| 57 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (91%), Xenopus tropicalis (83%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9NP61 | |
| ARFGAP3 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human ARFGAP3. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 26286 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction