missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARFGAP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551342
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ARFGAP2 protein.
ARFGAP2 gene ontology annotations related to this gene include GTPase activator activity; metal ion binding.
Specifications
| ARFGAP2 | |
| Polyclonal | |
| Unconjugated | |
| Arfgap2 | |
| 2310032E02Rik; ADP ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase activating protein 2; ADP-ribosylation factor GTPase-activating protein 2; ARF GAP 2; ARFGAP2; GTPase-activating protein ZNF289; IRZ; NBLA10535; Zfp289; Zinc finger protein 289; zinc finger protein 289, ID1 regulated; Znf289 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 362162, 77038, 84364 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.5 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q3MID3, Q8N6H7, Q99K28 | |
| Arfgap2 | |
| Synthetic peptide of human ARFGAP2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction