missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARFGAP1 Antibody (1C4E2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | ARFGAP1 |
|---|---|
| Clone | 1C4E2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18240504
|
Novus Biologicals
NBP2-61670 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633048
|
Novus Biologicals
NBP2-61670-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARFGAP1 Monoclonal specifically detects ARFGAP1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| ARFGAP1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| ADP-ribosylation factor 1 GTPase activating protein, ADP-ribosylation factor 1 GTPase-activating protein, ADP-ribosylation factor GTPase activating protein 1, ADP-ribosylation factor GTPase-activating protein 1, ARF GAP 1, ARF1 GAP, ARF1-directed GTPase-activating protein, ARF1GAP, bA261N11.3, FLJ10767, GAP protein, HRIHFB2281, MGC39924 | |
| ARFGAP1 | |
| IgG2b | |
| Protein G purified |
| 1C4E2 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Human, Mouse | |
| 55738 | |
| Purified recombinant fragment of human ARFGAP1 (AA: 270-414) expressed in E. Coli. | |
| Primary | |
| 44.7 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title