missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arfaptin 1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA549981
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Arfaptin 1 protein.
Arfaptin 1 Polyclonal antibody specifically detects Arfaptin 1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.
Specifications
| Arfaptin 1 | |
| Polyclonal | |
| Unconjugated | |
| ARFIP1 | |
| 2310030I10Rik; ADP ribosylation factor interacting protein 1; ADP-ribosylation factor interacting protein 1; ADP-ribosylation factor-interacting protein 1; Arfaptin; arfaptin 1; Arfaptin1; arfaptin-1; Arfip1; ARFP1; D3Ertd598e; HSU52521 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27236, 60382, 99889 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P53367, Q9JHU5 | |
| ARFIP1 | |
| Synthesized peptide derived from internal of human ARFIP1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction