missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARF6 Recombinant Rabbit Monoclonal Antibody (23GB3160), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | ARF6 |
|---|---|
| Clone | 23GB3160 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088720
![]() |
Invitrogen™
MA553390 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087751
|
Invitrogen™
MA553391 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARF6 Recombinant Monoclonal antibody specifically detects ARF6 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ARF6 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P62330, P62331, P62332 | |
| 11845, 382, 79121 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3160 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| ADP ribosylation factor 6; ADP-ribosylation factor 6; AI788669; ARF6; AW496366; DKFZp564M0264 | |
| A synthetic peptide of human ARF6 (120-175AA). | |
| Primary | |
| -20°C | |
| ARF6 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title