missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF6 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20225-1-AP-150UL
This item is not returnable.
View return policy
Description
The antibody is specific to ARF6.
ARF6 (ADP-ribosylation factor 6) is a broadly expressed, differentially regulated GTPase that is present in cytoplasm and on both cell-surface and intracellular membranes and whose functions may include tissue-specific effects on vesicular trafficking during cellular differentiation.Specifications
| ARF6 | |
| Polyclonal | |
| Unconjugated | |
| ARF6 | |
| ADP ribosylation factor 6, ARF6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11845, 382 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62330, P62331 | |
| ARF6 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction