missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11673-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody can bind ARFs for the close sequences.
ADP-ribosylation factor 1 is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I and class III, and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.Specifications
| ARF4 | |
| Polyclonal | |
| Unconjugated | |
| ARF4 | |
| ADP ribosylation factor 4, ARF2, ARF4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11843, 378, 79120 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18085, P61750, P61751 | |
| Arf4 | |
| ARF4 Fusion Protein Ag2272 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction