missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARF4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA536421
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 21 kDa. Purity is >95% by SDS-PAGE.
ADP-ribosylation factor 1 is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I and class III, and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| ARF4 | |
| Polyclonal | |
| Unconjugated | |
| ARF4 | |
| AA407803; ADP ribosylation factor 4; ADP-ribosylation factor 2; ADP-ribosylation factor 4; ARF2; Arf4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11843, 378 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P18085, P61750 | |
| ARF4 | |
| Synthetic peptide corresponding to amino acids 80-130 of human ARF4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction