missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARF4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537841
This item is not returnable.
View return policy
Description
This antibody is predicted to react with rat, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
ADP-ribosylation factor 1 is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I and class III, and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifications
| ARF4 | |
| Polyclonal | |
| Unconjugated | |
| ARF4 | |
| AA407803; ADP ribosylation factor 4; ADP-ribosylation factor 2; ADP-ribosylation factor 4; ARF2; Arf4 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 11843, 378 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P18085, P61750 | |
| ARF4 | |
| Peptide with sequence C-SEMTDKLGLQSLRN, from the internal region of ARF4 (aa137-150). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction