missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARF1 Mouse anti-Human, Mouse, Rat, Clone: ARF1/2117, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07774-100ug
This item is not returnable.
View return policy
Description
ARF1 Monoclonal specifically detects ARF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| ARF1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| ADP-ribosylation factor 1 | |
| Recombinant full-length human ARF1 protein | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| ARF1/2117 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 375 | |
| Store at 4°C. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction