missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARES™ Alexa Fluor™ 488 DNA Labeling Kit

Product Code. 10453432
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10453432 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10453432 Supplier Invitrogen™ Supplier No. A21665

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

ARES™ Alexa Fluor™ 488 DNA Labeling Kit

The ARES™ Alexa Fluor™ DNA Labeling Kit provides a versatile, two-step method for labeling DNA with our Alexa Fluor™ dyes. In the first step, an amine-modified nucleotide is incorporated into DNA using conventional enzymatic labeling methods. In the second step, the amine-modified DNA is chemically labeled using our proprietary amine-reactive Alexa Fluor™ 488 dye. The labeled probes can be used for fluorescence in situ hybridization (FISH) and microarray techniques. We offer ARES™ Alexa Fluor™ DNA Labeling Kits in five fluorescent colors, and each kit provides sufficient reagents for 5 to 10 labeling reactions of 1 to 5μg DNA each.

ARES™ Alexa Fluor™ Labeling Kit Specifications:
  • Dye (Ex/Em): Alexa Fluor™ 488 (492/520 nm)
  • Achieves more uniform, consistent labeling than techniques for enzymatic incorporation of labeled nucleotides
  • Typically produces one dye per 12–20 bases
  • Optimal for FISH and microarrays

    More Uniform Labeling With ARES™ Labeling Kits
    ARES™ Alexa Fluor™ Labeling Kits employ a two-step labeling technology—nick translation to enzymatically incorporate an amine-modified nucleotide (aminoallyl dUTP) followed by chemical labeling with Alexa Fluor™ dyes. This method achieves uniformity and consistency of labeling that is difficult to obtain with conventional enzymatic incorporation of labeled nucleotides.
    We also offer this two-step labeling technology in our FISH Tag™ DNA and FISH Tag™ RNA Kits, which provide a complete workflow solution for FISH applications, including all of the reagents for probe synthesis, labeling, purification and an anti-fade reagent to help protect the signal during fluorescence microscopy.

    More Options for Nucleic Acid Labeling
    To review various options for nucleic acid labeling (including DNA and RNA FISH), review Labeling Oligonucleotides and Nucleic Acids—Section 8.2 in the Molecular Probes™ Handbook.

    For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store in freezer (-5 to -30°C) and protect from light.
    Detection Method Fluorescence
    Format Kit
    Label or Dye Alexa Fluor 488
    Final Product Type Probes (Labeled DNA), cDNA (Labeled)
    Labeling Target DNA (General), cDNA
    Includes Label or Dye Yes
    Labeling Method Indirect Labeling
    Product Line ARES, Alexa Fluor
    Product Type DNA Labeling Kit
    Quantity 1 kit
    Shipping Condition Room Temperature
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.