missing translation for 'onlineSavingsMsg'
Learn More

Arenobufagin, MedChemExpress

Product Code. 16429158
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429158 10 mg 10mg
16421568 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16429158 Supplier Medchem Express Supplier No. HYN0876

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Arenobufagin is a natural bufadienolide from toad venom; has potent antineoplastic activity against HCC HepG2 cells as well as corresponding multidrug-resistant HepG2/ADM cells.

Arenobufagin is a natural bufadienolide from toad venom

Specifications

Chemical Name or Material Arenobufagin
CAS 464-74-4
Color White
Purity Grade Notes Research
Molecular Formula C24H32O6
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (240.09 mM; Need ultrasonic) ∣Ethanol : 10 mg/mL (24.01 mM; Need ultrasonic)
SMILES C[C@]([C@@H](C(C=C1)=COC1=O)CC2)(C3=O)[C@]2([C@@](CC[C@@]4([H])[C@@]5(CC[C@H](O)C4)C)([H])[C@]5([H])[C@@H]3O)O
Molecular Weight (g/mol) 416.51
Formula Weight 416.51
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.