missing translation for 'onlineSavingsMsg'
Learn More

Arecoline Hydrobromide, TRC

Product Code. 30138178
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138178 10 mg 10mg
30135995 500 mg 500mg
30009748 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138178 Supplier Toronto Research Chemicals Supplier No. TRCA76750010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 300-08-3
Molecular Formula C8 H13 N O2 . Br H
Molecular Weight (g/mol) 236.11
Synonym 3-Pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-methyl-, methyl ester, hydrobromide (1:1), 3-Pyridinecarboxylic acid, 1,2,5,6-tetrahydro-1-methyl-, methyl ester, hydrobromide (9CI), Nicotinic acid, 1,2,5,6-tetrahydro-1-methyl-, methyl ester, hydrobromide (8CI), Arecoline bromide, Arecoline hydrobromide, Methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, Methyl N-methyl-1,2,5,6-tetrahydronicotinate hydrobromide, Taeniolin
IUPAC Name methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide
SMILES Br.COC(=O)C1=CCCN(C)C1

Specifications

Analyte or Component Names Arecoline Hydrobromide
Shipping Condition Room Temperature
InChI Formula InChI=1S/C8H13NO2.BrH/c1-9-5-3-4-7(6-9)8(10)11-2;/h4H,3,5-6H2,1-2H3;1H
Solution Type Neat
Industry Type Forensics
Quantity 10 mg
Formula Weight 235.0208
Chemical Name or Material Arecoline Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.