missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Arctiin

Product Code. 16418966
Encompass_Preferred
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16418966 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16418966 Supplier MedChemExpress Supplier No. HYN0034

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Arctiin(NSC 315527), a plant lignan that can be extracted from the Arctium lappa (burdock) seeds, is a possible environmental endocrine disruptor compounds and have been shown to influence sex hormone metabolism as well as protein synthesis, steroid biosynthesis.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 534.55 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 20362-31-6
Product Type Natural Products
Purity 98.89%
Solubility DMSO : 250 mg/mL (467.68 mM∣Need ultrasonic) ∣H2O : 10 mg/mL (18.71 mM∣ultrasonic and warming and heat to 60°C)
Synonym Arctii∣ NSC 315527∣ Arctigenin-4-glucoside
Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.