missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Arctigenin

Product Code. 16498828
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16498828 Supplier MedChemExpress Supplier No. HYN0035
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Arctigenin ((-)-Arctigenin), a biologically active lignan, can be used as an antitumor agent. Arctigenin exhibits potent antioxidant, anti-inflammatory and antiviral (influenza A virus) activities. Arctigenin can be used for the research of metabolic disorders, and central nervous system dysfunctions.

Specifications

Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 372.41 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 7770-78-7
Product Type Natural Products
Purity 98.46%
Solubility DMSO : ≥ 125 mg/mL (335.65 mM)
Synonym (-)-Arctigenin
Target MMP∣Influenza Virus∣Autophagy∣Apoptosis
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.