missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Arctigenin

Product Code. 16428966
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16428966 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16428966 Supplier MedChemExpress Supplier No. HYN0035
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Arctigenin ((-)-Arctigenin), a biologically active lignan, can be used as an antitumor agent. Arctigenin exhibits potent antioxidant, anti-inflammatory and antiviral (influenza A virus) activities. Arctigenin can be used for the research of metabolic disorders, and central nervous system dysfunctions.

Specifications

Content And Storage Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 372.41 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 7770-78-7
Product Type Natural Products
Purity 98.46%
Solubility DMSO : ≥ 125 mg/mL (335.65 mM)
Synonym (-)-Arctigenin
Target MMP∣Influenza Virus∣Autophagy∣Apoptosis
Quantity 100 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.