missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Arctic Gloves
Description
- Grain goatskin.
- Water repellant Scotchguard™ treatment.
- Synthetic fur.
- Elasticized on back with zig-zag stitching.
- Good flexibility and outstanding dexterity.
- Supple.
- Good mechanical resistance.
Specifications
Specifications
| Certifications/Compliance | CE CATEGORY: II Intermediary risks, EN 420-2003 EEC Basic standards, EN 388-2003 Mechanical hazards, EN 511-2004 Cold hazards, EC Category PPE: 2, Quality Assurance: ISO 9001/2000, EC Certificate Number: N°0075/797/162/07/12/0587, EC Attestation Number: N°0075/797/162/07/12/0587 |
| Classification | Dexterity: 5 |
| Color | Beige |
| Cut Resistance Rating | 2 |
| Material | Leather |
| Product Type | Gloves |
| Size | 11 |
| For Use With (Application) | Agriculture, Building, Construction, Catering, Energy, Electricity, Food, Green Spaces, Iron, Steel, Logistics, Offshore, Ship Building, Telecom, Utility and Wood industries |
| Includes | Elasticized back, Scotchguard™ treatment |
| Liner Material | Synthetic fur |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?