Learn More
Epredia™ Arcos™ Trays and Storage
Product Code. 16292272 Shop All Epredia Products
Click to view available options
Quantity:
Set of four wheels
Product Type:
Wheels (Set of 4)
Product Code. 16292272
Brand: Epredia™ B84710009
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Enhance traceability and organization with trays, storage cabinets and cardboard sleeves for the Epredia™ Arcos™ block management system.
Smart and Secure Storage
  • The Arcos system offers flexible archiving solutions to fit customer needs for both short and long-term archiving. The Arcos storage cabinets offer two configurations with 18 and 24 tray capacity. These are designed specifically for high-density storage that can be stacked to optimize capacity and space.
  • For short-term archiving, the cabinets can be placed in the main lab area so the blocks can be accessed with ease. The 18 tray cabinets can be stacked three-units high, and the 24 tray cabinets can be stacked two-units high.
  • For long-term archiving, cardboard sleeves may be used as an alternative storage option in place of the Arcos cabinets. Each sleeve can hold one Arcos tray, up to 240 blocks.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wheels (Set of 4)
For Use With (Equipment) 18- and 24-tray Epredia™ Arcos™ cabinets
Quantity Set of four wheels
Epredia™ Arcos™ instrumentation requires the use of Arcos trays, sold separately. Arcos cabinets are highly recommended for storage.