Learn More
STEMCELL Technologies ArciTect™ High-Fidelity DNA Polymerase Kit
Polymerase, buffers, and dNTPs for high-fidelity PCR amplification
Brand: STEMCELL Technologies 76026
Additional Details : Weight : 0.00220kg
Description
- ArciTect™ High-Fidelity DNA Polymerase Kit is optimized for robust, high-fidelity DNA amplification (> 50X higher than Taq)
- It may be used in applications requiring ultra-low error rates, such as detection of genome editing with ArciTect™ T7 Endonuclease I (refer to the Technical Bulletin: Evaluation of Genome Editing), as well as sequencing, cloning/subcloning, synthetic biology, and SNP analysis
- ArciTect™ High-Fidelity DNA Polymerase is a fusion protein with a double-stranded DNA-binding domain and a Pyrococcus-like proofreading polymerase domain
Specifications
Pluripotent Stem Cells | |
1 Kit |