missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies ArciTect™ Cas9 Nuclease

Product Code. 17119301
Encompass_Preferred
Click to view available options
Quantity:
100 μg
300 μg
Unit Size:
100µg
300µg
This item is not returnable. View return policy

Product Code. 17119301

Brand: STEMCELL Technologies 76002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cas9 nuclease (4 μg/μL) for the generation of double-strand breaks in CRISPR-Cas9 genome editing

  • ArciTect™ Cas9 Nuclease requires association with a guide RNA—e.g. ArciTect™ sgRNA (Catalog #200-0013) or a duplex composed of ArciTect™ tracrRNA (Catalog #76016) and ArciTect™ crRNA (Catalog #76010)—to form a ribonucleoprotein (RNP) complex
  • This RNP complex then creates double-strand breaks at site-specific locations in the genome
  • ArciTect™ Cas9 Nuclease contains a nuclear localization signal at the N-terminus, ensuring that the RNP complex translocates to the nucleus, thereby increasing the efficiency of genome editing
  • As the RNP complex is fully functional upon transfection, it allows for immediate activity following translocation to the nucleus
  • The RNP complex is degraded over 48 hours, allowing sufficient time for genome editing to occur while reducing off-target effects that can be caused by the continuous presence of the RNP complex
  • The ArciTect™ Cas9 nuclease has been validated for use with human PSCs, T cells, hematopoietic stem & progenitor cells, and intestinal organoids

Specifications

Quantity 100 μg
Cell Type Pluripotent Stem Cells
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.