Learn More
Manutan Archive boxes, white, 267mm x 290mm x 387mm
Description
Excellent for storing, organising and recording documents. Suitable for A4 papers, ring binders and document wallets. Archive boxes can be easily stacked and used on racking. Complete with removable lid allowing easy access to all stored records. Made from up to 90% recycled material. Hand holes for easy carrying.
- White
- Capacity : 10kg
- Dimensions (H x W x D) : 267mm x 290mm x 387mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.