missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARC/NOL3 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB500-257AF647
This item is not returnable.
View return policy
Description
ARC/NOL3 Polyclonal antibody specifically detects ARC/NOL3 in Human samples. It is validated for Western Blot
Specifications
| ARC/NOL3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| Apoptosis repressor with CARD, ARCFLJ35304, CARD2, Muscle-enriched cytoplasmic protein, MYC, MYP, NOP30, NOPnucleolar protein 3, nucleolar protein 3 (apoptosis repressor with CARD domain), Nucleolar protein of 30 kDa | |
| A synthetic peptide made to residues 125-141 (PELQTLTRPGALRAPRR) of the human Nop30 protein. | |
| 0.1 mL | |
| Apoptosis | |
| 8996 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction