missing translation for 'onlineSavingsMsg'
Learn More

Arbutin, TRC

Product Code. 30348399
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30348399 25 mg 25mg
30344207 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30348399 Supplier Toronto Research Chemicals Supplier No. TRCA76651025MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Arbutin
CAS 497-76-7
Purity Grade Notes HPLC
Molecular Formula C12 H16 O7
Quantity 25 mg
Synonym Arbutin, 4-Hydroxyphenyl β-D-glucopyranoside, Arbutine, Arbutoside, Arbutyne, Hydroquinone glucose, Hydroquinone β-D-glucopyranoside, NSC 4036, Ursin, Uvasol, p-Arbutin, p-Hydroxyphenyl β-D-glucopyranoside, p-Hydroxyphenyl β-D-glucoside, β-Arbutin
InChI Formula InChI=1S/C12H16O7/c13-5-8-9(15)10(16)11(17)12(19-8)18-7-3-1-6(14)2-4-7/h1-4,8-17H,5H2/t8-,9-,10+,11-,12-/m1/s1
SMILES OC[C@H]1O[C@@H](Oc2ccc(O)cc2)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-hydroxyphenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol) 272.25
Formula Weight 272.0896
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.