missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Arbutin

Product Code. 16468888
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16468888 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16468888 Supplier MedChemExpress Supplier No. HYN0192
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Arbutin (β-Arbutin) is a competitive inhibitor of tyrosinase in melanocytes, with Kiapp values of 1.42 mM for monophenolase; 0.9 mM for diphenolase. Arbutin is also used as depigmenting agents. Arbutin is a natural polyphenol isolated from the bearberry plant Arctostaphylos uvaursi, possesses with anti-oxidant, anti-inflammatory and anti-tumor properties.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 272.25 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 497-76-7
Product Type Natural Products
Purity 98.0%
Solubility DMSO : ≥ 50 mg/mL (183.65 mM) ∣H2O : 33.33 mg/mL (122.42 mM∣Need ultrasonic)
Synonym β-Arbutin
Target Tyrosinase
Quantity 10 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.