Learn More
Honeywell™ ARATHERMA NOMEX™ Heavy Gloves
Product Code. 12665333
Click to view available options
:
Size: 7
Product Code. 12665333
Brand: Honeywell™ 223207007
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Honeywell™ ARATHERMA NOMEX™ Heavy Gloves for protection against thermal hazards up to 250°C.
  • 100% Nomex™ seamless glove
  • Lined with a seamless
  • The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
  • The glove offers maximum comfort and added sturdiness
  • The cotton interlock lining strengthens the glove's mechanical performance while absorbing perspiration
  • The glove is ambidextrous and thus more cost-effective
  • The glove's relative thickness (gauge 13) enables it to absorb shocks, making it suitable for the handling of heavy objects
  • The NOMEX™ fiber pills only a little and ensures good heat resistance for removing dishes from the oven up to 250°C
  • White in color, it is compatible with the agrifood industry while enhancing hygiene and cleanliness
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-03 Mechanical Hazards, EN 407-04 Thermical hazards
Description 13 gauge
Color White
Hand Ambidextrous
Material Nomex, Cotton
Size 7
Length (Metric) 26.5 cm