missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ ARATHERMA FIT SHORT Grip Gloves
Click to view available options
:
Size: 10
Description
- 100% knitted cotton glove
- Double face coating of nitrile dots
- Ambidextrous
- The knitting technique helps to obtain a glove that conforms snugly to the hand with good dexterity
- The glove offers maximum comfort and added sturdiness
- The glove is ambidextrous and thus more cost-effective
- The cotton lining reinforces the glove's mechanical performance while absorbing hand perspiration for added comfort
- The double face coating in nitrile brings a good grip and is more econmic as ambidextrous
- The Cotton increases the glove's cutting resistance and thermal resistance (contact heat)
- The quality of the materials used creates an effective barrier against heavy and sharp parts
Specifications
Specifications
| Certifications/Compliance | EN 420-2003 EEC Basic standard, EN 388-2016 Mechanical hazards, EN 407-2004 Thermical hazards |
| Description | 10 gauge |
| Material | Cotton |
| Size | 10 |
| Length (Metric) | 25 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction