Learn More
Honeywell™ ARATHERMA FIT SHORT Gloves
Description
- Knitted heavy para-aramid (Twaron™ or Kevlar™)
- Lined with one cotton knitted glove
- The knitting technique helps to obtain a glove that conforms snugly to the hand and good dexterity
- The glove offers maximum comfort and added sturdiness
- Kevlar™ glove lined with cotton is pleasant to the touch
- The lining enhances the glove's mechanical performance while absorbing perspiration
- The glove is ambidextrous and thus more cost-effective
- The glove's relative thickness (gauge 7) enables it to absorb shocks, making it suitable for the handling of heavy objects
- Good resistance to cuts and protection against thermal hazards
- The cotton liner increase the thermal resistance
Specifications
Specifications
| Certifications/Compliance | EN 420-03 : EEC Basic standards, EN 388-03 : Mechanical hazards, EN 407-04 : Thermical hazards |
| Description | 7 gauge |
| Material | Para-Aramid |
| Size | 9 |
| Liner Material | Cotton |
| Length (Metric) | 26 cm |