missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARAP1 Monoclonal Antibody (ARAP1-2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119728
This item is not returnable.
View return policy
Description
This antibody reacts with C-terminal part of ARAP1 (intracellular epitope), a 160 kDa adaptor protein.
ARAP1 (angiotensin II type 1 receptor-associated protein) is an adaptor protein with ArfGAP and RhoGAP activities, containing five PH domains and ankyrin repeat. This adaptor seems to serve as a link between phosphoinositide- Arf-, and Rho-mediated cell signaling. ARAP1 supports the plasma membrane recycling of angiotensin II type 1 receptor (AT1) and is important also for cell-specific trafficking of pro-death receptor TRAIL-R1 (DR4) to the plasma membrane, thus promoting the TRAIL-induced apoptosis in certain cell types. ARAP1 also affects cell spreading.
Specifications
| ARAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| Q96P48 | |
| Arap1 | |
| His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| ARAP1-2 | |
| Unconjugated | |
| Arap1 | |
| 2410002L19Rik; arap1; Arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1; ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1; ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containing protein 1; centaurin, delta 2; centaurin-delta-2; Centd2; cnt-d2; Kiaa0782; mKIAA0782 | |
| Mouse | |
| Protein A | |
| RUO | |
| 116985 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction