missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARAP1 Antibody (ARAP1-2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68801
This item is not returnable.
View return policy
Description
ARAP1 Monoclonal specifically detects ARAP1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| ARAP1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:50 | |
| ARAP1 | |
| His6-tagged protein corresponding to amino acids 1190-1450 of human ARAP1 | |
| Protein A purified | |
| RUO | |
| Primary | |
| The ARAP1-2 reacts with C-terminal part of ARAP1, a 160 kDa adaptor protein. | |
| Store at 4C. Do not freeze. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| ARAP1-2 | |
| Unconjugated | |
| arf-GAP with Rho-GAP domain, ANK repeat and PH domain-containing protein 1, ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 1, centaurin, delta 2, Centaurin-delta-2, CENTD2ARF-GAP, RHO-GAP, ankyrin repeat, and pleckstrin homology domains-containingprotein 1, cnt-d2, KIAA0782centaurin-delta-2 | |
| Mouse | |
| 160 kDa | |
| 0.1 mg | |
| Cell Cycle and Replication | |
| 116985 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction