missing translation for 'onlineSavingsMsg'
Learn More

Araldite Rapid Epoxy Resin Adhesive and Hardener

Product Code. 12715298
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Ceramics, Metal, Glass, Wood
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
12715298 Ceramics, Metal, Glass, Wood Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12715298 Supplier Araldite Rapid Supplier No. AA/R

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
An epoxy resin and hardener in separate tubes. Drying time 10min to 15min. Suitable for use with ceramics, metal, glass and wood.

Specifications

Product Type Epoxy Resin Adhesive and Hardener
Format Paste
For Use With (Application) Suitable for use with ceramics, metal, glass and wood
For Use With (Equipment) Ceramics, Metal, Glass, Wood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.