missing translation for 'onlineSavingsMsg'
Learn More

Arachidonic acid, MedChemExpress

Product Code. 16417037
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16417037

Brand: Medchem Express HY109590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Arachidonic acid is an essential fatty acid and a major constituent of biomembranes.

Arachidonic acid is an essential fatty acid and a major constituent of biomembranes.

Specifications

Chemical Name or Material Arachidonic acid
Physical Form Liquid
CAS 506-32-1
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H32O2
For Use With (Application) COVID-19-immunoregulation
Synonym Immunocytophyt
Solubility Information Ethanol : 100 mg/mL (328.44 mM; Need ultrasonic) ∣DMSO : ≥ 50 mg/mL (164.22 mM)
SMILES CCCCC/C=C\C/C=C\C/C=C\C/C=C\CCCC(O)=O
Molecular Weight (g/mol) 304.47
Formula Weight 304.47
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.